VWR® Funnels, PTFE
Supplier: VWR International
89026-140 89026-142 89026-146 89026-148 89026-144
89026-140EA 421.22 CAD
89026-140 89026-142 89026-146 89026-148 89026-144
VWR® Funnels, PTFE
Funnels Filtration Funnels Filter Funnels
Isostatically molded, FDA approved pure PTFE molding provides a durable, heavy wall construction with a smooth, non-stick internal finish.
  • Chemically inert
  • May be used from 0 to 280 °C (32 to 536 °F)
Suitable for a variety of laboratory applications.