Funnels for FLASH Chromatography, California Pacific Lab, CP Lab Safety
Supplier: CP Lab Safety
CF-960WN CF-3005 CF-60 CF-120 CF-240 CF-960
33501-900EA 103.87 CAD
33501-900 47735-396 33501-892 33501-894 33501-896 33501-898
Funnels for FLASH Chromatography, California Pacific Lab, CP Lab Safety
Funnels Filtration Funnels Filter Funnels
Ideal for use in FLASH chromatography applications
Made of unbreakable polypropylene. Funnels feature a closed-end design that facilitates the gentle addition of a solvent to the column. The funnel stem is perforated to permit solvent to be poured down the solvent wall. The insertion of the funnel into the column will not disturb the top layer of sand or silica gel.