Super Erecta®/Super Adjustable Super Erecta® Top-Track™ High-Density Storage Systems, Metro™
Supplier: Metropolitan Wire
LTTA18C LTTA21C LTTA24C LTTE18C LTTE21C LTTE24C LTTM18C LTTM21C LTTM24C TTA18C TTA18K3 TTA18S TTA21C TTA21K3 TTA21S TTA24C TTA24K3 TTA24S TTE18C TTE18S TTE18K3 TTE21C TTE21K3 TTE21S TTE24C TTE24K3 TTE24S TTM18C TTM18S TTM18K3 TTM21C TTM21S TTM21K3 TTM24C TTM24K3
CA83009-472EA 2842.87 CAD
CA83009-472 CA83009-474 CA83009-476 CA83009-478 CA83009-480 CA83009-482 CA83009-484 CA83009-486 CA83009-488 CA83009-518 CA83009-520 CA83009-522 CA83009-524 CA83009-526 CA83009-528 CA83009-530 CA83009-532 CA83009-534 CA83009-536 CA83009-538 CA83009-540 CA83009-542 CA83009-544 CA83009-546 CA83009-548 CA83009-550 CA83009-552 CA83009-554 CA83009-556 CA83009-558 CA83009-560 CA83009-562 CA83009-564 CA83009-566 CA83009-568
Super Erecta®/Super Adjustable Super Erecta® Top-Track™ High-Density Storage Systems, Metro™
Furniture Shelves
Top-Track™ system increases storage capacity up to 50% and maximizes the usable storage area in a given space.
  • Chrome-plated units are ideal for dry environments with minimal exposure to corrosives
  • Type 304 stainless steel units provide maximum protection in corrosive environments
Metroseal 3™ units are made of electroplated carbon steel with a unique corrosion-resistant epoxy, an economical alternative to stainless steel units, and are coated with antimicrobial product protection which inhibits the growth of bacteria, mold, mildew, and fungi thatMore Product Information