Autoclavable Shelving and Accessories, Metro™
Supplier: Metropolitan Wire
9986S LC5S1860 LC5S2460 LC1860AC LC2460AC 5MHTP 5MHTPB 5MHTN 5MHTNB 33UPS-SW 54UPS-SW 63UPS-SW
CA82002-502EA 55.18 CAD
CA82002-502 CA97002-832 CA97002-834 CA97002-836 CA97002-838 CA97002-852 CA97002-854 CA97002-856 CA97002-858 CA97002-862 CA97002-864 CA97002-866
Autoclavable Shelving and Accessories, Metro™
Furniture Shelves
Preconfigured shelving is ideal for autoclaving cages and other equipment
Autoclavable covers are rated for approximately 70 cycles. Casters have a load rating of 136 kg (300 lbs.) and are temperature rated to 149 °C (300 °F).
Ordering information: Cage racks come with aluminum split sleeves and swedged posts. Autoclavable casters must be ordered separately.