Diamond RealSeal™ Plastic Jerricans
Supplier: Globe Scientific
7190005 7190010 7190020
77448-002EA 106.87 CAD
77448-002 77448-004 77448-006
Diamond RealSeal™ Plastic Jerricans
Bottles Jerricans
Provide efficient storage, simple transport, and easy dispensing of large liquid volumes in and around the laboratory.
  • Produced from High-Density Polyethylene (HDPE)
  • Safe for freezer storage down to −100 °C
  • Resistant to most corrosives and suitable for a wide variety of laboratory uses
Rugged and durable high-density polyethylene construction make Diamond RealSeal™ jerricans highly resistant to chemicals, rustproof and molded graduations provide approximate volumes in liters and gallons.
Rectangular design and low center of gravity provide efficient and stable liquid storage, while the angled spout and molded-in handle facilitates easy pouring.
Jerricans are precision molded from virgin resins for consistent wall thickness and smooth interior and exterior surfaces. The jerricans include leak-proof polypropylene caps that are engineered with an integral seal ring for leak proof closure.