Stacking, Hanging, Interlocking Polypropylene Bins, 10⁷/₈" Depth
Supplier: Triton Products
3-235B 3-235BK 3-235GR 3-235GRN 3-235R 3-235Y
77269-506PK 141.55 CAD
77269-506 77269-508 77269-512 77269-514 77269-516 77269-520
Stacking, Hanging, Interlocking Polypropylene Bins, 10⁷/₈" Depth
Boxes Dispenser Boxes
LocBin® is the only bin that can stack, hang, lock and nest to maximize space.
  • Unique dual interlocking channel system slides and locks bins together guaranteeing bins will not collapse, cave in or topple into one another
  • Mount to wall using the 5 to 600 wall mount rail system, on Louvered panels or stack on a flat surface to provide multiple storage options for small items
  • Bottom interlocking rail allows stacking bins to be nested offering the user a smaller footprint in tight spaces
  • Extra deep groove system accepts bin dividers that stay in place when
More Product Information