Boston Round Bottles, Clear, Narrow Mouth, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5110120V-21 5110220V-21 5110422V-21 5110824V-21 5111628V-21 5113233V-21 5110120C-21 5110422C-21 5110824C-21 5111628C-21 5113233C-21 5111628V-22 5113233V-22 5111628V-24 5113233V-24 5110120C-24 5110220C-24 5110422C-24 5110824C-24 5111628C-24 5110120V-25 5110220V-25 5110422V-25 5110824V-25 5111628V-25 5113233V-25 5110120C-25 5110220C-25 5110422C-25 5110824C-25 5111628C-25 5113233C-25 5110120V-26 5110220V-26 5110422V-26 5110824V-26 5111628V-26 5113233V-26 5110422C-26 5110120B 5110220B 5110422B 5110824B 5111628B 5113233B
89045-104CS 315.37 CAD
89045-104 89045-106 89045-108 89045-110 89045-112 89045-114 89045-116 89045-118 89045-120 89045-122 89045-124 89045-126 89045-136 89045-138 89045-184 89045-186 89045-188 89045-190 89045-192 89045-194 89045-196 89045-198 89045-200 89045-202 89045-204 89045-206 89045-208 89045-210 89045-212 89045-214 89045-216 89045-218 89045-220 89045-222 89045-224 89045-226 89045-228 89045-230 89045-232 89045-234 89045-240 89045-248 89045-250 89045-252 89045-254 89045-256 89045-258
Boston Round Bottles, Clear, Narrow Mouth, Kimble®, DWK Life Sciences
Bottles Glass Bottles
High quality, economical narrow mouth bottles are manufactured from clear Type III soda-lime glass to ensure easy viewing of the contents.
Supplied with black phenolic caps.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are also packaged in bulk packs. Pallet packs consist of shrink-wrapped glasswareMore Product Information