Super Erecta® Wire Shelving, Chrome-Plated, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta®
1824NC 2454NC 2142NC 1860NC 1448NC 2130NC 2130NC 2424NC 2442NC 1854NC 2436NC 2136NC 3672NC 1424NC 1872NC 1860NC 3060NC 1830NC 1842NC 2160NC 2424NC 3072NC 2154NC 1442NC 2160NC 2124NC 1836NC 2172NC 2436NC 3648NC 1848NC 1448NC 1472NC 2154NC 2472NC 2442NC 2172NC 2448NC 1824NC 1818NC 1436NC 1842NC 2148NC 1836NC 1460NC 2454NC 1872NC 1460NC 3660NC 3060NC 3036NC 3672NC 1436NC 3048NC 2148NC 1442NC 1430NC 2430NC 1818NC 2460NC 1830NC 3636NC 2460NC 2142NC 1472NC 3072NC 3036NC 1430NC 3660NC 2448NC 2136NC 1848NC 3048NC 1424NC 3636NC 3648NC 2430NC 1854NC 2124NC 2472NC
CA22233-178EA 420.21 CAD
CA22233-178 CA19766-094 19766-162 19766-082 19766-080 CA22233-176 CA19766-090 CA19766-454 19766-400 CA19766-212 CA19766-134 CA19766-452 19766-200 CA19766-176 19766-364 19766-166 19766-086 CA19766-416 19766-208 CA19766-414 CA19766-456 CA19766-136 CA19766-334 CA19766-082 19766-192 CA19766-080 19766-070 19766-158 19766-114 CA19766-200 CA19766-166 CA19766-364 19766-234 CA19766-086 19766-196 CA19766-162 19766-076 19766-230 CA19766-208 19766-118 CA19766-400 CA19766-192 CA19766-070 19766-182 19766-620 CA19766-234 19766-466 19766-464 CA19766-076 CA19766-230 CA19766-196 19766-142 19766-428 CA19766-118 22221-498 CA19766-158 CA19766-114 19766-106 CA19766-182 19766-094 22233-178 22233-176 19766-090 19766-136 19766-334 19766-456 CA19766-464 19766-134 CA22221-498 19766-454 19766-212 CA19766-142 19766-176 19766-452 CA19766-428 CA19766-106 19766-416 CA19766-620 19766-414 CA19766-466
Super Erecta® Wire Shelving, Chrome-Plated, Metro™
Furniture Shelves
Standard shelves up to 121.9 cm (48") in length can support up to 363 kg (800 lbs.) per level. Standard shelves longer than 121.9 cm (48") and all super wide shelves can support up to 272 kg (600 lbs.) per level.
  • Chrome-plated shelves are perfect for dry environments with minimal exposure to corrosives
  • Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items
Note: The 35.6 cm (14") wide shelves require the use of footplates (see 19766-814 series) when used as a freestanding unit. The actual length of the shelves is 3.2 mm (⅛") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. Stationary posts over 220 cm high are not for use with shelves less than 61 cm (24") wide.
Easy shelf adjustment is possible through the use of SiteSelect® stationary or mobile posts (sold separately), which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each shelf and post is sold separately. To order posts or accessories, contact your Avantor representative for more information. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts.