Two Pegboards, 18-Gauge Steel Square Hole with Mounting Hardware
Supplier: Triton Products
LB18-G LB18-W LB18-B LB18-BK
77269-652PK 321.99 CAD
77269-652 77269-660 77269-642 77269-644
Two Pegboards, 18-Gauge Steel Square Hole with Mounting Hardware
Furniture Backsplashes, Wallracks, Wallpanels
LocBoard® is made from 18 gauge steel and offers the ultimate holding capacity for the most demanding tool and storage needs. It is 12 times stronger than conventional pegboard.
  • Ideal for heavy, oversized and odd shaped professional power tools, hand, maintenance tools and more
  • Two boards can be mounted vertically or horizontally together or positioned on multiple wall areas as required
  • Use with LocHook® 50000 Series 100% heavy-duty steel 2 and 4-point contact locking hooks
  • All hooks lock securely on the board and are easily repositioned
  • Mounting hardware is hidden from sight - presenting a clean, attractive and durable appearance
  • 350 lbs. load rating per board
LocBoard® heavy-duty epoxy coated steel pegboards with square holes offer the ultimate holding capacity for the most demanding tool and storage needs. The epoxy coating makes them scratch resistant. LocBoard® heavy-duty steel pegboard is ideal for use in a clean room, factory floor production area, maintenance, repair areas, home and garage. Use only LocHook® 50000 Series industrial strength hooks and steel shelf with patented 2 and 4-point contact locking mechanisms to create the most secure and reliable pegboard system for commercial and home tool storage and other work items. The most demanding tool storage requirements for power tools, large and odd shaped tools are easily met using the LocBoard steel pegboards.
Ordering information: Limited lifetime warranty.
Delivery information: Mounting hardware included.