Stacking, Hanging, Interlocking Polypropylene Bins, 14³/₄" Depth
Supplier: Triton Products
3-240B 3-240BK 3-240GR 3-240GRN 3-240R 3-240Y
77269-524PK 187.2 CAD
77269-524 77269-526 77269-530 77269-532 77269-534 77269-538
Stacking, Hanging, Interlocking Polypropylene Bins, 14³/₄" Depth
Boxes Dispenser Boxes
LocBin® is the only bin that can stack, hang, lock and nest to maximize space.
  • Unique dual interlocking channel system slides and locks bins together guaranteeing bins will not collapse, cave in or topple into one another
  • Mount to wall using the 5-600 wall mount rail system, on louvered panels or stack on a flat surface to provide multiple storage options for small items
  • Bottom interlocking rail allows stacking bins to be nested offering the user a smaller footprint in tight spaces
  • Extra deep groove system accepts bin dividers that stay in place when
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable