LocHook® Double Rod Zinc Plated Steel Pegboard Hooks for LocBoard®
Supplier: Triton Products
52628 52113 52119 52213 52313 52319 52419
77269-256PK 52.54 CAD
77269-256 77269-244 77269-246 77269-248 77269-250 77269-252 77269-254
LocHook® Double Rod Zinc Plated Steel Pegboard Hooks for LocBoard®
Wall Hooks
The patented LocHook® provides the ultimate building block system for secure organization and storage.
  • Ideal for storing hammers, sledge hammers, large hand tools and power tools when used with other LocHooks
  • The 2 and 4-point contact locking mechanism offers industrial strength holding capability
  • Hooks mount vertically and horizontally in 4 distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
  • Fixtures install from the front, so no need to remove the board for installation or repositioning of LocHooks® causing no damage to the LocBoard®
  • Compatible with all LocBoard® sizes and other brands of steel tool boards with square holes spaced 1 to ¹/₂" O.C.
Having secure, commercial grade, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. LocHook®(s) and LocBoard® present an ideal steel pegboard tool storage solution for companies that support 5S and Lean Manufacturing initiatives. They provide the most heavy-duty pegboard and hook system and are ideal for dependable storage for factory floor, shop, home and garage. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Together, the LocBoard and LocHook will provide the perfect steel pegboard solution. Imagine a clean looking commercial grade pegboard with the strongest hooks available to hang your items within grasp at all times. That is the power of the steel pegboard system from Triton Products.
Ordering information: Limited lifetime warranty.