Super Erecta Shelf® Solid Shelving, All-Stainless Steel, Metro™
Supplier: Metropolitan Wire
1424NFS 1430NFS 1436NFS 1442NFS 1448NFS 1460NFS 1824NFS 1830NFS 1836NFS 1842NFS 1848NFS 2124NFS 2130NFS 2136NFS 2142NFS 2148NFS 2160NFS 2424NFS 2430NFS 2436NFS 2442NFS 2448NFS 2460NFS
CA98000-680EA 961.41 CAD
CA98000-680 CA98000-682 CA98000-684 CA98000-686 CA98000-688 CA98000-690 CA98000-692 CA98000-694 CA98000-696 CA98000-698 CA98000-700 CA98000-702 CA98000-704 CA98000-706 CA98000-708 CA98000-710 CA98000-712 CA98000-714 CA98000-716 CA98000-718 CA98000-720 CA98000-722 CA98000-724 CA98000-726
Super Erecta Shelf® Solid Shelving, All-Stainless Steel, Metro™
Furniture Shelves
Solid shelving is available in four widths and six lengths, all stainless solid shelf with stainless corners.
  • All-stainless, 18-gauge steel construction with ¹/₈" (3 mm) ship's edge detail helps contain spills
  • All-stainless design, including stainless cast corners, can be used in cart wash and autoclave environments
Shelves up to 121.9 cm (48") in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") can support up to 272 kg (600 lbs.) per level.
Note: The 35.6 cm (14") wide shelves require the use of footplates (More Product Information