VWR® Light Meter
Supplier: VWR
VWR®
3252
21800-014EA 729.03 CAD
21800-014
VWR® Light Meter
Light Meters
Dual range light meter reads both Lux and Foot Candles on 3.5 cm (13/8") LCD.
  • Traceable®
  • Dual range and display
Unit features zero reference settings for tungsten, fluorescent, daylight, and mercury light sources
Data acquisition system (61161-386) captures, displays, and stores readings on any PC. Information can be imported into databases. Includes 31/2" diskette and 91.4 cm (36") serial cable with D9F plug.
Displays percent differential from a reference point and recalls highest, lowest, and average readings. A “Data Hold” button freezes the display to capture readings. Precise photo diode and color correction filter assist in providing cosine and color corrected measurements. Serial RS-232 output allows meter to be connected to a computer or data logger (21800-026) for monitoring and storing results.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Meter is supplied with photo cell, 101.6 cm (40") cable, serial computer output, and 9V battery.