Super Erecta® Wire Shelving, Stainless Steel, Metro™
Supplier: Metropolitan Wire
1424NS 1430NS 1448NS 1460NS 1472NS 1824NS 1848NS 1860NS 2148NS 2460NS 1854NS 1436NS 1442NS 1830NS 1836NS 1842NS 1872NS 2124NS 2130NS 2136NS 2142NS 2154NS 2160NS 2172NS 2424NS 2430NS 2436NS 2442NS 2448NS 2454NS 2472NS 3636NS 3660NS 3672NS
CA19766-072EA 671.19 CAD
CA19766-072 CA19766-074 CA19766-084 CA19766-088 CA19766-092 CA19766-098 CA19766-122 CA19766-140 CA19766-168 CA19766-204 CA19766-386 CA19766-504 CA19766-506 CA19766-508 CA19766-510 CA19766-512 CA19766-516 CA19766-518 CA19766-520 CA19766-522 CA19766-524 CA19766-526 CA19766-528 CA19766-530 CA19766-532 CA19766-534 CA19766-536 CA19766-538 CA19766-540 CA19766-542 CA19766-544 CA19767-056 CA19767-060 CA19767-062
Super Erecta® Wire Shelving, Stainless Steel, Metro™
Furniture Shelves
Type 304 stainless steel shelves provide maximum protection in corrosive environments.
  • Open-wire design
  • Easy shelf adjustment
  • Double grooves to aid shelf alignment
Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items
Shelves up to 121.9 cm (48) in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") can support up to 272kg (600lbs.) per level.
Note: The 35.6 cm (14) wide shelves require the use of footplates (see 19766-814 series) whenMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable