Metro® Super Adjustable™ Wire Shelving, Chrome Plated, Metro™
Supplier: Metro International
Super Adjustable 2™ Super Erecta Shelf® Super Adjustable™
A1448NC A1430NC A2172NC A1836NC A2136NC A2448NC A3072NC A2172NC A2436NC A3048NC A2148NC A2472NC A1460NC A1436NC A2460NC A2130NC A2154NC A2448NC A2436NC A3660NC A2460NC A1448NC A1472NC A1860NC A2160NC A3648NC A1436NC A2142NC A3036NC A2430NC A2148NC A2430NC A2442NC A1836NC A2154NC A2136NC A1842NC A1424NC A3072NC A1442NC A2160NC A1430NC A1830NC A2424NC A2124NC A3636NC A2454NC A2142NC A3672NC A2442NC A3060NC A1860NC A2472NC A1442NC A1824NC A1854NC A1460NC A2124NC A1842NC A1824NC A1872NC A1848NC A1424NC A1472NC A3648NC A1854NC A3048NC A2130NC A1872NC A3672NC A2454NC A3660NC A2424NC A1848NC A3636NC A1830NC A3060NC A3036NC
CA22233-730EA 456.58 CAD
CA22233-730 22233-720 89369-790 CA22233-734 22233-724 CA22233-732 22233-722 89369-832 89369-754 89369-796 CA89369-760 89369-872 CA89369-838 89369-826 89369-868 CA89369-832 CA89369-754 22233-718 CA22233-726 22233-716 CA22233-724 CA22233-728 CA22233-722 CA22233-720 89369-766 CA89369-852 CA89369-772 89369-760 CA89369-848 89369-838 CA89369-802 CA89369-844 CA89369-766 CA22233-716 CA89369-808 CA22233-718 22233-742 22233-740 22233-746 22233-744 CA89369-784 89369-852 CA89369-860 89369-772 CA89369-814 89369-848 CA89369-856 CA89369-778 89369-802 89369-844 22233-738 CA22233-746 89369-808 22233-732 CA22233-740 22233-730 22233-736 CA22233-744 22233-734 CA22233-742 89369-820 89369-864 CA89369-796 CA89369-872 89369-784 89369-860 CA89369-790 CA89369-826 CA89369-868 89369-814 89369-856 89369-778 CA89369-820 CA89369-864 CA22233-738 22233-728 CA22233-736 22233-726
Metro® Super Adjustable™ Wire Shelving, Chrome Plated, Metro™
Furniture Shelves
Durable chrome-plated shelves are ideal for dry environments with minimal exposure to corrosives.
  • Design wire shelf system with appropriate accessories
An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Notes: The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. The 245.4 cm high stationary posts are not for use with shelves less than 61 cm wide.
Adjustable shelves feature a unique corner release system that allows for quick and easy shelf adjustment without the use of tools. Shelves are designed for use with SiteSelect® stationary or mobile posts, which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the posts.
Certifications: Units meet MIL-S-40144E specifications.
Ordering information: Each shelf and post are sold separately. All shelves include four pairs of plastic split sleeves for securing shelf to posts.