Super Erecta Shelf* Solid Shelving, Standard Stainless Steel, Metro*
Supplier: Metro International
Super Erecta® Super Erecta Shelf®
1836LS 1848FS 2442LS 2424LS 1848LS 2442FS 1442LS 2148LS 1448LS 1442LS 2160FS 1424FS 1836FS 2436LS 2136LS 2124LS 2124LS 2448FS 1824LS 1448FS 2460LS 1460LS 1860FS 2430LS 2424FS 1842LS 1424LS 2442FS 2448LS 1860LS 1830FS 1830FS 2460FS 1848FS 1842LS 2136LS 2430FS 1430LS 2160FS 1830LS 2136FS 2142LS 1860LS 2142FS 2142LS 2436FS 1460FS 2130LS 2130LS 2130FS 1430FS 1436LS 2448LS 2448FS 1460FS 2424LS 1448FS 2160LS 2460FS 2148FS 2442LS 1824LS 2436LS 1842FS 2148FS 1848LS 2124FS 1824FS 2142FS 1842FS 1430LS 1824FS 1424LS 2424FS 2436FS 2460LS 1448LS 1442FS 1436FS 1436LS 2160LS 1442FS 1430FS 1460LS 1830LS 2136FS 2430LS 1836FS 2148LS 1860FS 2124FS 2130FS 1836LS 1424FS 2430FS 1436FS
CA97005-650EA 1803.45 CAD
97005-280 97005-282 97005-284 CA97005-650 97005-640 97005-720 CA97005-652 97005-840 97005-642 CA97005-720 97005-634 97005-832 CA97005-840 CA97005-642 97005-710 97005-636 CA97005-722 97005-834 CA97005-644 97005-712 97005-836 CA97005-724 97005-638 97005-714 CA97005-800 CA97005-646 CA97005-726 97005-838 CA97005-802 97005-716 CA97005-648 CA97005-728 97005-718 CA97005-280 CA97005-282 CA97005-284 97005-630 97005-632 97005-830 CA97005-640 CA97005-632 CA97005-830 CA97005-832 CA97005-634 CA97005-710 97005-822 CA97005-636 CA97005-834 97005-704 CA97005-712 97005-824 97005-706 CA97005-836 CA97005-638 97005-826 CA97005-714 97005-628 97005-708 CA97005-838 97005-828 CA97005-716 CA97005-718 97005-740 CA97005-630 97005-732 97005-734 CA97005-822 97005-736 CA97005-704 CA97005-824 97005-738 CA97005-706 CA97005-826 CA97005-628 CA97005-708 CA97005-828 97005-650 97005-652 97005-654 CA97005-740 97005-730 97005-722 CA97005-654 CA97005-730 97005-644 97005-724 97005-800 97005-646 CA97005-732 97005-726 CA97005-734 97005-802 97005-648 97005-728 CA97005-736 CA97005-738
Super Erecta Shelf* Solid Shelving, Standard Stainless Steel, Metro*
Furniture Shelves
Solid shelving features 18-gauge stainless steel construction with flat, embossed, or louvered/embossed surfaces and black powder-coated corners.
  • Ship's edges help contain spillage minimize contamination and permit easier cleanup
  • Adjustable feet (bolt levelers) compensate for surface irregularities
Two-fold thickness forms a 3.2 mm (¹/₈") raised ship's edge on all four sides for all-around rigidity and high strength. Embossed or louvered/embossed shelf designs allow air circulation under packages and containers. Shelves can be loaded and unloaded from all sides for maximum use of storage space.
Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. The 245.4 cm high stationary posts are not for use with shelves less than 61 cm wide.
Sturdy aluminum castings firmly lock the corners to the posts (sold separately). Each post has rolled, circular grooves along its entire length at 2.5 cm (1") intervals. Unique tapered split sleeve (plastic or aluminum) snaps together around each post. Tapered openings in the shelf corners slide over the tapered split sleeves, providing a positive lock. Shelf is assembled in minutes without the use of nuts, bolts, or special tools.
Ordering information: Stability decreases as the ratio of height to width increases; units should be kept as wide and low as possible. Each shelf and post is sold separately. For casters used with mobile units, see 19766-236 series. For special posts used to create shelving units with counter tops, see 19766-814 series. For other accessories, contact your Avantor representative.