Boston Round Bottles, Vacuum and Ionized, Amber, Narrow Mouth, Qorpak
Supplier: Qorpak
GLC-01906 GLC-01928 GLC-01950 GLC-01979 GLC-02006 GLC-01976 GLC-07017 GLC-07102 GLC-07104 GLC-02031 GLC-10283 GLC-07548 GLC-01911 GLC-07149 GLC-09176 GLC-02011
11311-184CS 2572.02 CAD
11311-184 11311-454 36319-725 36319-726 36319-727 36319-769 36319-770 36319-771 36319-772 36319-773 93000-946 93000-948 89204-844 89237-492 30617-142 93000-945 10820-604 10820-606 76487-258 76487-260 76487-262 76487-264 76487-266 76487-268 76487-270 76487-272 76487-274 76487-276 76487-278 76487-280
Boston Round Bottles, Vacuum and Ionized, Amber, Narrow Mouth, Qorpak
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Boston round bottles are made of glass
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with your product in any way.