Super Erecta® Work-in-Process Shelving, Metro™
Supplier: Metropolitan Wire
SF35N4S SF36N4S
CA19766-802EA 557.16 CAD
CA19766-802 CA19766-804 CA19766-806
Super Erecta® Work-in-Process Shelving, Metro™
Furniture Shelves
Super Erecta® rod top shelves with 5 cm (2") slope support wafer boats and carriers.
  • Shelf capacity: 90.7 kg (200 lbs.)
  • Open wire construction maintains laminar air flow
  • Electropolished Type 304 stainless steel construction
The use of an open four-sided shelf frame will help increase stability when the lowest position shelf is greater than 30.5 cm (12") above the floor.
Easy shelf adjustment is possible through the use of SiteSelect® stationary or mobile posts, which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. The height of the stationary posts includes a leveling bolt and cap.
Nonconductive, but can be made conductive with the addition of aluminum split sleeves (see 19766-814 series).