Boston Round Bottles, Amber, Narrow Mouth
Supplier: Qorpak
GLC-07386 GLA-00895 GLA-00902 GLA-00890 GLC-06420 GLC-02030 GLC-04927 GLA-00884 GLC-02032 GLC-01918 GLC-01939 GLC-01967 GLC-01992 GLC-01982 GLC-02018 GLC-02001 GLC-01937 GLC-01990 GLC-02015 GLC-01912 GLC-01935 GLC-01961 GLC-01985 GLC-02012 GLC-01904 GLC-01926 GLC-01947 GLC-01977 GLC-02004 GLA-00896 GLA-00900 GLC-01905 GLC-01927 GLC-01971 GLC-01922 GLC-05639 GLC-01997 GLC-02010 GLC-01915 GLA-00885 GLC-01898 GLA-00886 GLC-01903 GLC-01909 GLC-01960 GLA-00893 239190 239191 239193 239195 239196 272128 GLC-01921 GLC-01970
11311-102CS 321.73 CAD
11311-102 11311-228 11311-230 11311-590 15712-408 15712-412 15712-416 15712-418 15712-420 15712-424 15712-436 16054-108 16054-112 16054-134 16085-296 16087-296 16087-298 16087-318 16087-320 16150-953 16152-020 16152-041 16152-063 16152-085 16152-096 16152-097 16152-099 16152-100 16152-110 16152-223 16152-245 16152-267 16152-289 16152-405 16152-427 16152-449 16152-460 16152-482 16152-642 16152-686 16152-700 16152-722 16153-102 16153-135 16153-168 16153-190 16153-226 16218-222 16218-224 36319-393 36319-395 36319-410 36319-411 36319-414 36319-435 93000-938 93001-642 16152-098 16152-201 89215-522 89237-486 89237-488 89237-490 30617-126 30617-130 30617-132 30617-134 30617-138 30617-140 89423-618 15712-431 16152-076 16218-221 76170-310 76170-312 76170-314 76170-316 76170-318 76210-028 76419-710 76419-712 76419-718 76419-720 76419-724 76419-726 76419-732 76419-734 76419-736 76419-740 76419-742 76472-440 76487-182 76487-184 76487-186
Boston Round Bottles, Amber, Narrow Mouth
Bottles Glass Bottles
Amber Boston round bottles protect contents from UV rays and are ideal for light sensitive products.
  • Round bottles are made of glass
These general use bottles are perfect for liquids. They feature a round body, rounded shoulders, and narrow screw neck opening. These environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage.
Ordering information: Bottles 36319-393, 36319-395, 93000-908, 93000-938, and 82004-068 are supplied pre-cleaned for volatiles. Bottle 16054-134 is supplied pre-cleaned for semi-volatiles. Bottle 89215-522 is supplied ultra cleaned. Bottles 11311-405, 10820-604, and 93000-945 are supplied vaccum and ionized. Bottles 16152-740, 16218-217, and 16218-219 are supplied without caps. Bottles 76487-182, 76487-184 and 76487-186 are cleaned and certified for semi-volatiles.