Boston Round Bottles, Amber, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216836 W216837 W216838 W216839 W216840 W216842 W216843 W216844 W216845 W216846 W216847 W216848 W216849 W216850 W216851 W216852 W216853 W216854 W216855 W216856 W216857 W216858 W216859 W216860 W216861 W216862 W216863 W216864 W216865 W216866 W216867 W216868 W216869 W216870 W216841
14235-684CS 120.42 CAD
14235-684 14235-686 14235-688 14235-690 14235-692 14235-696 14235-698 14235-700 14235-702 14235-704 14235-706 14235-708 14235-710 14235-712 14235-714 14235-716 14235-718 14235-720 14235-722 14235-724 14235-726 14235-728 14235-730 14235-732 14235-734 14235-736 14235-738 14235-740 14235-742 14235-744 14235-746 14235-748 14235-750 14235-752 14235-694
Boston Round Bottles, Amber, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton Boston round bottles are manufactured from amber soda-lime glass that conforms to USP Type III requirements.
Boston round bottles are offered with a choice of PTFE faced foam polyethylene liner with polypropylene cap, poly-vinyl liner with polypropylene cap, PE cone liner with black phenolic cap, or rubber liner with black phenolic cap.
These narrow mouth bottles facilitate pouring and storage of liquids. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. The partitioned carton protects bottles from scratches during shipping, andMore Product Information