Boston Round Bottles, Clear, Narrow Mouth
Supplier: Qorpak
GLC-06419 GLC-06421 GLC-01159 GLC-01189 GLC-01176 GLC-01209 GLA-00805 GLC-01094 GLC-01092 GLC-01116 GLC-01114 GLC-01106 GLC-01138 GLC-01173 GLC-01205 GLC-01204 237403 GLC-01226 GLC-01237 GLC-01082 GLC-01099 GLC-01123 GLC-01151 GLC-01185 GLC-01217 GLC-01091 GLC-01112 GLC-01135 GLC-01170 GLC-01203 GLC-01234 GLA-00809 GLA-00811 GLA-00815 GLC-01089 GLC-01109 GLC-01131 GLC-01166 GLC-01196 GLC-01228 GLC-01087 GLC-01128 GLC-01162 GLC-01191 GLC-01221 GLC-01081 GLC-01098 GLC-01181 GLC-01193 GLC-01894 GLA-00807 GLA-00817 GLA-00816 GLC-01143 GLC-01211 GLC-01178 GLC-01111 GLC-01153 GLC-01101 GLA-00819 GLC-01164 GLC-01134 GLC-04870 GLC-01146 GLC-01171 GLC-06413 GLA-00812 239202 239203 239204 239205 239206 239207 272134 272133 272130 272129 272126 272123 272118 GLC-01142 GLA-00806 GLA-09761
11311-644CS 939.72 CAD
11311-644 15712-410 15712-414 15712-422 15712-426 15712-434 16054-102 16054-104 16087-006 16151-027 16151-038 16151-049 16151-050 16151-051 16151-060 16151-082 16151-107 16151-118 16151-120 16151-124 16151-129 16151-220 16151-242 16151-286 16151-300 16151-322 16151-344 16151-435 16151-457 16151-490 16151-515 16151-537 16151-559 16151-610 16151-612 16151-614 16151-650 16151-672 16151-719 16151-730 16151-752 16151-774 16151-865 16151-887 16151-901 16151-923 16151-945 16151-967 16152-018 16152-116 16152-124 16152-126 16152-203 16152-211 16192-057 16193-901 16218-090 16218-207 16218-210 16218-212 36319-125 36319-212 36319-236 36319-238 36319-241 36319-245 82027-146 93001-592 93001-626 93001-628 93001-636 93001-638 93001-654 16151-592 89204-852 89204-856 89204-862 89204-864 89204-866 89204-868 89204-904 76170-320 76170-322 76170-324 76170-326 76170-328 76170-330 76210-026 76419-706 76419-708 76419-714 76419-716 76419-722 76419-728 76419-730 76419-738 76487-188 76487-190 76539-346 76539-358
Boston Round Bottles, Clear, Narrow Mouth
Bottles Glass Bottles
Clear, flint glass Boston round bottles offer maximum visibility and sample integrity.
  • USP certified Type III (soda lime) glass
These general use bottles are perfect for liquids, product storage, and field or plant sampling projects. They feature a round body, rounded shoulders, and narrow screw neck opening. Environmentally sensitive bottles help to eliminate waste and help to ensure product integrity for long term storage.
Ordering information: Bottles 16152-203, 16152-209, 16152-211, 11311-644, 93001-592, 93001-626, 93001-628, 93001-636, and 93001-638 are supplied Ultra Cleaned. Bottles 36319-125, 36319-212, and 36319-245 are supplied Cleaned for Volatiles. Bottle 36319-140 is supplied Cleaned and Certified for Volatiles. Bottle 93001-654 is supplied Cleaned and Certified for Semi-Volatiles.