Centrifuge Bottles
Supplier: Globe Scientific
6330 6332
76363-2410CS 150.88 CAD
76363-2410 76363-2412
Centrifuge Bottles
Bottles Centrifuge Bottles
Rugged polypropylene centrifuge bottles are suitable for low to moderate speed centrifugation when filed to 80% of their nominal capacity.
  • Consistent wall thickness for low to moderate speed centrifugation
  • Polypropylene (PP) bottle and cap with integral seal ring for leakproof closure
  • Bottle must be filled to 80% of volume for proper performance
  • Reusable and autoclavable at 121 °C
  • Excellent chemical resistance
Reusable and autoclavable, these bottles are designed with leakproof linerless closures. Bottles are molded with consistent wall thickness for excellent performance and offer excellent chemical resistance.