SP Bel-Art Sterile Cloning Cylinders, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37847-0300 F37847-0100 F37847-0200 H37847-0000
47727-256CS 321.53 CAD
47727-256 77776-774 77776-776 62866-140
SP Bel-Art Sterile Cloning Cylinders, Bel-Art Products, a part of SP
Cylinders Cloning Cylinders
Cloning cylinders improve colony isolation by assuring a homogenous culture of cells rather than a mixed population.
  • Polystyrene construction
  • Sterile
  • Assure a homogenous culture of cells
  • Cylinders are tapered in shape for wider bottom
To isolate incubated colonies, a cloning cylinder is placed around the colony of choice and attached by sterile grease. After adding trypsin into the cylinder, the dish is placed into an incubator until cells detach. Cells are then collected and transferred to another vessel. Cylinders are tapered so the bottom edge is wider than the top to afford a good seal. Molded of polystyrene.
Ordering information: Individual sizes includes 5 sterile bags of 10. Assorted pack includes 4 sterile bags of 10 of each size, 12 bags total.