KIMAX® Funnels, Long Stem, 60° Angle, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
28980-150 28980-200 28980-250
89001-414CS 260.98 CAD
89001-414 30214-149 30214-160
KIMAX® Funnels, Long Stem, 60° Angle, Kimble Chase, DWK Life Sciences
Funnels Filtration Funnels Filter Funnels
Constructed with heavy uniform walls and strong, fire polished rims and stems for a long service life
These funnels are made of low extractable borosilicate glasses.
Funnels have a high resistance to chemical attack, mechanical shock, and thermal shock.
Certifications: Designed from ASTM Specification E1095, Type I, Class B requirements.