Synthware Round-Bottom Flasks with Hooks, Kemtech America
Supplier: Kemtech America
F331405 F331410 F331425 F331450 F339100
60002-392EA 47.28 CAD
60002-392 60002-396 60002-400 60002-404 60002-408
Synthware Round-Bottom Flasks with Hooks, Kemtech America
Flasks Round Bottom Flasks
Heavy wall design.
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various joint sizes available
  • Hooks for retention springs
This round bottom flask features hooks to secure the connection using retention springs.