Corning® Spinner Flasks with Vertical Sidearms, Corning
Supplier: Corning
4510-8L 4510-15L 4510-36L 4512-8L 4512-15L 4512-36L
22877-128CS 3141.59 CAD
22877-128 22877-130 22877-132 22877-134 22877-136 22877-138
Corning® Spinner Flasks with Vertical Sidearms, Corning
Flasks Spinner Flasks
Borosilicate glass flasks are baffled to enhance agitation of contents.
  • Side baffles enhance aeration and agitation of flask contents
  • Proprietary 'stretch' impeller design ensures optimal stirring
  • Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.
Accessories information: Proculture® spinner direct top drive motor with high torque, low rpmMore Product Information