Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2110-0002 2110-0006 2110-0008 2110-0016 2110-0032
16120-950CS 273.02 CAD
16120-950 16120-953 16120-956 16120-959 16120-962
Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Bottles Roller Bottles
Square, polypropylene wide mouth bottles are designed to replace glass serum and general-purpose storage bottles.
  • Resistant to acids, alcohols, and bases
  • Contact-clear bottles provide visible liquid level
  • Wide mouth for easy cleaning, sample filling, and retrieval
  • Square shape saves space and offers large labeling surface
  • Leakproof and repeatedly autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.