PYREX® Volumetric Flasks, Screw Cap, Class A, Corning
Supplier: Corning
5650-100 5650-25 5650-50
89091-064CS 1214.07 CAD
89091-064 89091-262 89091-438
PYREX® Volumetric Flasks, Screw Cap, Class A, Corning
Flasks Volumetric Flasks
Made of PYREX® borosilicate glass.
Flasks have an expanded open end to facilitate transfer of crystals, salts and powders. Standard GPI thread screw cap has an inert, cone-shaped, polyethylene liner for leakproof closure. Conforms to Class A capacity tolerances.