Nalgene® Large Sample Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2120-0005 2120-0010
16125-118CS 240.16 CAD
16125-118 16125-129
Nalgene® Large Sample Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Widely used for collecting specimens and water sampling.
  • Leakproof
  • With polypropylene screw closure
Also excellent for use with dry chemicals. Mouth opening: 89 mm (3¹/₂").