TUNAIR™ Shake Flasks, IBI Scientific
Supplier: IBI Scientific
SS-4001 SS-4002 SS-4003 SS-8001 SS-8002 SS-8003 SS-5011 SS-5012 SS-5013 SS-2002C SS-2003C SS-6002C SS-6002S SS-6003C SS-6003S SS-2001-S SS-2002S SS-2003S SS-2001C SS-6001C SS-6001S SS-2012 SS-2013
14217-566CS 160.91 CAD
14217-566 14217-568 14217-570 14217-572 14217-574 14217-576 82031-188 82031-190 82031-192 95025-498 95025-500 95025-502 95025-504 95025-506 95025-508 IBSS-2001S IBSS-2002S IBSS-2003S 89235-056 89235-058 89235-060 89235-062 89235-064
TUNAIR™ Shake Flasks, IBI Scientific
Flasks Cell Culture Flasks
TUNAIR™ Shake Flask Systems are a unique and patented flask and closure system designed for microbiology and biotechnology applications
The two piece cap assemblies are constructed of polypropylene and are resistant to most solvents. All caps and flasks are fully autoclavable and the filter linings can be autoclaved or replaced. All TUNAIR™ filter linings are 0.22 micron and are available in silicone or nitrocellulose. TUNAIR™ caps are available in four convenient colors: red, blue, green, and yellow. The working volume of the 300ml flask is 100ml and for the 2.5L flask is 1L. The shakerMore Product Information