Oblong Bottles, High-Density Polyethylene, Wide Mouth
Supplier: Qorpak
PLC-09005 PLC-09004 PLC-07195 PLA-03259 PLA-03252 PLA-03254 PLC-03467 PLC-03469 PLC-03472 PLC-03473 PLC-03494 PLC-03476 PLC-03477 PLC-03481 PLC-03482 PLC-03487 PLC-03488 PLC-03478 PLA-03253 PLC-03475 PLC-03479
11311-580CS 1268.78 CAD
11311-580 11311-596 11311-598 11311-632 16087-130 16087-184 16087-186 16125-976 16125-984 16125-985 16125-986 16125-987 16125-988 16125-989 16125-990 16125-991 16125-992 16125-993 16125-996 16125-997 16196-299 16196-302 82027-770 76539-374 76539-376 76539-378
Oblong Bottles, High-Density Polyethylene, Wide Mouth
Bottles Roller Bottles
Natural HDPE wide mouth Oblong bottles offer a space saving design for the workbench.
  • Bottles feature a wide mouth opening, ideal for sampling solids and powders
  • HDPE construction provides an excellent moisture barrier and reliable impact resistance
They are ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping.
Ordering information: Bottles 16196-299, 16196-302, and 16125-976 are supplied pre-cleaned for volatiles. Bottles 11311-580, 11311-596, 11311-598, and 11311-600 are supplied Vacuumed and Ionized.