MiraWIPE® Wipers, Foamtec International
Supplier: Foamtec International
HT5740B HT5790 HT5790B HT5790S HT5790S-25
89066-272CS 4837.99 CAD
89066-272 10810-938 10810-940 10810-942 89069-040
MiraWIPE® Wipers, Foamtec International
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Constructed from continuous filament microfiber polyester, woven wipers feature superior absorbency and contamination pick-up when compared to traditional knitted polyester wipers.
Woven, abrasion-resistant fabric minimizes particle generation from wiper face and edge.
Ordering information: Cleanroom laundered and packaged. Wipers are available in configurations to meet ISO Class 3 to 6 requirements; contact your Avantor sales representative for more information.