UltraSOLV® Chamber Cleaning Pads, Wipers, and Swabs, Foamtec International
Supplier: Foamtec International
HT4500-TXX2 HT4536D-10 HT4513PD-10 HT1702-50 HT4790 HT4540D-10 HT4520PA-10 HT4580A-10 HT4510-10 HT1701-5 HT1700-5 HT1732-50 HT4754 HT4512P-10 HT4580-10 HT4560-10 HT4540-10 HT4528-10 HT4522-10 HT4518D-10 HT4528D-10 HT4580D-10 HT4536DS-10
89068-844EA 549.16 CAD
89068-844 10810-816 10810-818 10810-907 10810-936 10810-952 10810-954 10810-960 10810-998 10811-751 10811-753 10811-759 10811-769 10811-771 10811-773 10811-775 10811-777 10811-779 10811-781 10811-783 10811-785 10811-787 77776-834
UltraSOLV® Chamber Cleaning Pads, Wipers, and Swabs, Foamtec International
Wipes Cleaning Kits and Pads
Tools are designed to work together to clean recessed surfaces and microporous sub-surfaces without using hazardous solvents. When used in conjunction, they safely and quickly remove build-up on process equipment and reduce contamination levels. Choose from diamond, silicon carbide, or aluminum oxide ScrubPADS for initial scouring. Complete the cleaning process with hydrophilic polyurethane foam sponges, wipers, and swabs.
  • Use ScrubPADS for initial scouring
  • Complete cleaning process with foam sponges, wipers, and swabs
ScrubPADS are fully launderedMore Product Information