VWR® Wide Mouth Bottles, Rectangular Amber HDPE
Supplier: VWR International
414004-196 414004-197 414004-198 414004-199 414004-200
414004-196CS 455.93 CAD
414004-196 414004-197 414004-198 414004-199 414004-200
VWR® Wide Mouth Bottles, Rectangular Amber HDPE
Bottles Plastic Bottles
Space-saving, leakproof amber bottles are for sampling, storage, or shipment of light-sensitive dry chemicals and liquids.
  • HDPE construction
  • Amber tint protects against UV radiation
  • Ideal for holding light-sensitive dry chemicals and liquids
Bottles comply with current USP requirements for maximum light transmission. Sturdy construction prevents bulging, translucent material allows for optical clarity, and wide mouth makes filling easy. Bottles have excellent chemical resistance to most acids, bases, and alcohols, and can be stored in freezer to −100°More Product Information