VWR® Wide Mouth Bottles, Rectangular, HDPE,
Supplier: VWR International
414004-191 414004-192 414004-193 414004-194 414004-195
414004-191CS 299.59 CAD
414004-191 414004-192 414004-193 414004-194 414004-195
VWR® Wide Mouth Bottles, Rectangular, HDPE,
Bottles Plastic Bottles
Space-saving, leakproof bottles are for sampling, storage, or shipment of dry chemicals or liquids.
  • Sturdy construction
  • Space-saving design
  • Leakproof
  • HDPE fabrication
Sturdy construction prevents bulging, translucent material allows for optical clarity, and wide mouth makes filling easy. Bottles have excellent chemical resistance to most acids, bases, and alcohols, and can be stored in freezer to −100 °C (−148 °F). Bottle 414004-195 features integrated handle for better grip, greater safety, and pouring convenience. Polypropylene screw caps included.
Order Now
SPECIFICATIONS
Environmentally Preferable