Glow™ and Universal Gel Loading Dyes, G-Biosciences
Supplier: G-Biosciences
786-100
786-101
786-102
786-103
786-104
786-105
786-106
786-107
CA82022-462EA
205.35
CAD
CA82022-462
CA82022-464
CA82022-466
CA82022-468
CA82022-470
CA82022-472
CA82022-474
CA82022-476
Glow™ and Universal Gel Loading Dyes, G-Biosciences
Stains
Electrophoresis Stains
Premixed and ready-to-use gel dyes are designed to minimize the use of ethidium bromide
They are specifically formulated for nucleic acid electrophoresis. Glow dyes provide intense DNA/RNA bands with little background or band distortion on any type of agarose or acrylamide gel. Universal Loading Dyes™ are Ficoll®-based and do not contain any ethidium bromide. Both RNA loading dyes are formamide-based. Glow RNA is ideal for quick screenings of RNA preps.