VWR® Wide Mouth Laboratory Bottles, Amber HDPE
Supplier: VWR International
414004-116 414004-117 414004-118 414004-119 414004-120 414004-121 581300 581310 581320 581330 581340 581350
414004-116CS 546.57 CAD
414004-116 414004-117 414004-118 414004-119 414004-120 414004-121 89221-670 89221-672 89221-674 89221-676 89221-678 89221-680
VWR® Wide Mouth Laboratory Bottles, Amber HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use, but are especially good for light-sensitive materials or photo- sensitive samples.
  • Ideal for light-sensitive materials or photosensitive samples
  • Leakproof
  • Easy-to-pour wide mouth design
Can be stored in freezer to −100 °C (−148 °F), and have excellent chemical resistance to alcohols, akalis, and both diluted and concetrated acids. Thick walls afford durability. Shrink ring at neck ensures smooth interior so that contents pour fluidly, while sealing ring and semi-buttressed threads in closure ensure leak prevention. Bottles are produced via the injection blow molding process for exact features and bottle stability.
Ordering information: Amber polypropylene caps included.