VWR® Wide Mouth Laboratory Bottles, LDPE
Supplier: VWR International
414004-104 414004-105 414004-106 414004-107 414004-108 414004-109
414004-104CS 325.24 CAD
414004-104 414004-105 414004-106 414004-107 414004-108 414004-109
VWR® Wide Mouth Laboratory Bottles, LDPE
Bottles Plastic Bottles
Translucent, flexible bottles are for general laboratory use, and are especially good for shipping liquids and for trace metal analysis.
  • Ideal for shipping liquids and for trace metal analysis
  • Impact-resistant
  • Leakproof
  • Easy-to-pour wide mouth design
Can be stored in freezer to −100 °C (−148 °F). Offer better visibility of contents than high-density polyethylene bottles, plus excellent chemical resistance to alcohols, alkalis, and both diluted and concentrated acids. Thick walls afford durability, yet bottles are still squeezable. Shrink ring at neckMore Product Information