KIMAX® Reverse Scale Graduated Cylinders, Class A, To Deliver, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20028W-10 20028W-100 20028W-1000 20028W-2000 20028W-25 20028W-250 20028W-50 20028W-500
89001-682CS 452.01 CAD
89001-682 89001-684 89001-686 89001-688 89001-690 89001-692 89001-694 89001-696
KIMAX® Reverse Scale Graduated Cylinders, Class A, To Deliver, Kimble Chase
Cylinders Measuring Cylinders
Cylinders are made of sturdy borosilicate glass and feature reverse metric graduations in white
Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.