Lab Style Bottles, High-Density Polyethylene, Wide Mouth, Qorpak®
Supplier: Qorpak
PLA-03173 PLA-03175 PLA-03177 PLA-03168 PLA-03170 PLA-03172 PLA-03174 PLA-03176 PLA-03178 PLA-03169
11310-986CS 1014.1 CAD
11310-986 11310-988 11310-990 11311-136 11311-140 16086-068 16086-072 16086-076 16086-080 16086-086 16086-090 82024-072
Lab Style Bottles, High-Density Polyethylene, Wide Mouth, Qorpak®
Bottles Plastic Bottles
Wide mouth HDPE lab style bottles are supplied with linerless polypropylene caps.
  • Ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping
Excellent chemical resistance and the leak resistant closure make these ideal for storing, shipping, and packaging liquids for long periods. Linerless white polypropylene caps provide good impact strength, cost effectiveness, and pliability.
Packaging: Caps are attached on the convenience pack and enclosed on the bulk pack.