KIMAX® Single Metric Scale Graduated Cylinders, Class B, with Polyethylene Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20039P-100 20039P-50 20039P-1000
89000-766CS 3562.07 CAD
89000-766 89001-214 89001-216
KIMAX® Single Metric Scale Graduated Cylinders, Class B, with Polyethylene Stoppers, Kimble Chase
Cylinders Measuring Cylinders
Calibrated to contain
Cylinders are designed from ASTM Specification E1272, Style 2, Class B requirements.
Cylinders are made of sturdy borosilicate glass. Scale is durable white ceramic enamel. Yellow and blue [ST] stopper is made from linear high-density polyethylene and has a closed bottom. The enlarged top of the stopper protects the neck if the cylinder is knocked over.
Ordering information: Replacement stoppers are available; see the 89000-797 series.