SP Bel-Art Precisionware™ Bottles, Polypropylene, Wide Mouth, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F10632-0005 F10632-0006 F10632-0007 F10632-0008
89005-736CS 6501.87 CAD
89005-736 74950-021 74950-023 74950-025 74950-027
SP Bel-Art Precisionware™ Bottles, Polypropylene, Wide Mouth, Bel-Art Products, a part of SP
Bottles Roller Bottles
For pharmaceutical or similar packaging of liquid or dry materials.
  • Polypropylene construction
  • Deep, square threads
  • Smooth, rounded interiors
  • Easy to clean and thoroughly empty
  • Autoclavable
Polypropylene bottles are Boston-round style, clear, and molded from FDA compliant material. Polypropylene caps have a valve-seal design for a leakproof fit. Deep, square threads help prevent back-off during shipping.
Ribbed cap exteriors are designed for torque closure and easy opening. Smooth, rounded bottle interiors are designed for easy cleaning and thorough emptying. Autoclavable, with caps detached, to 121 °C (250 °F).