KIMAX® Graduated Cylinders, Class A, To Deliver, Serialized and Certified, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20026-10 20026-100 20026-1000 20026-25 20026-250 20026-50 20026-500
89001-666CS 1872.49 CAD
89001-666 89001-668 89001-670 89001-672 89001-674 89001-676 89001-678
KIMAX® Graduated Cylinders, Class A, To Deliver, Serialized and Certified, Kimble Chase
Cylinders Measuring Cylinders
Cylinders are made of sturdy borosilicate glass with a hexagonal glass base that has been flat ground for stability.
Cylinders feature a single metric scale
Designed from ASTM Specification E1272, Style 1, Class A requirements. Each cylinder is permanently marked with an individual serial number and is supplied with a Certificate of Graduation Accuracy.
The 10mL cylinder has an enlarged funnel-shaped top for ease of filling. Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.