Nunc® MAX-100 CryoStore™ Boxes, Thermo Scientific
Supplier: Thermo Fisher Scientific
330821 374187 341483 374187
CA55710-250CS 1201.27 CAD
CA55710-250 CA55710-252 CA55710-254 55710-252
Nunc® MAX-100 CryoStore™ Boxes, Thermo Scientific
Boxes Cryoboxes
Stackable, impact-resistant boxes are made from molded polycarbonate and fit most rack storage systems
The MicroMAX-100 box stores up to 64 microcentrifuge 1.5mL tubes with caps. The MAX-100 box permits storage of 100 internally threaded 1.0–1.8mL tubes. The MegaMAX-100 box permits storage of 100 internally threaded 3.6mL tubes. Divider accessories are made of polyethylene-coated cardboard.
Provide 23% more storage than many boxes of comparable size. Boxes feature a printed grid on top for easy identification of contents, and the dividers can be removed completely for storage of larger containers.
Ordering information: The MicroMAX-100 box includes an 8x8 divider. MAX-100 and MegaMax-100 boxes include a removable 10x10 divider, which can be replaced with an optional 9x9 divider to accommodate externally threaded vials. The MegaMAX-100 box used with the 9x9 divider can also accommodate vials sealed in CryoFlex™ tubing. For the optional 9x9 divider that fits the MAX-100 box, contact your representative.