VWR® Cylinder Guards
Supplier: VWR International
15001-374 15001-376 15001-378 15001-380 15001-382 15001-384
15001-374PK 91.31 CAD
15001-374 15001-376 15001-378 15001-380 15001-382 15001-384
VWR® Cylinder Guards
Supports Support Sleeves
Made of sponge rubber in a hexagonal shape to prevent rolling.
  • Hexagonal shape prevents rolling
Center hole is sized for standard graduated cylinders.