Diamond® Essentials™ Griffin Beakers, with Handle
Supplier: Globe Scientific
3654-250 3656-1M 3656-250 3656-2M 3656-600
77447-914EA 9.01 CAD
77447-914 77447-924 77447-926 77447-928 77447-932
Diamond® Essentials™ Griffin Beakers, with Handle
Beakers
These Griffin style beakers meet the demands for mixing, pouring, stirring, and measuring solutions in many laboratory operations.
  • Manufactured from virgin, USP Class VI resins
  • Bold, printed, dual scale graduations
  • Single non-drip spout
  • Autoclavable at 121 °C
  • Chemically resistant to most acids, bases, and many common solvents
  • Beakers without handles stack to save storage space
These beakers are the safe, shatterproof alternative to glass in operations where plastic is suitable. Value, quality, and performance are insured by precision molding in an automated manufacturing process. These beakers are designed with handles that facilitate easy pouring.
Available in economic polypropylene (PP) or crystal clear polymethylpentene (PMP).
Certifications: Complies with FDA 21-CFR and European Pharmacopeia.