Men's Flame-Resistant/Chemical Splash Protection Lab Coats
Supplier: Bulwark Protection
KNR6RB-2L-0R KNR6RB-3L-0R KNR6RB-4L-0R KNR6RB-5L-0R KNR6RB-LG-0R KNR6RB-MD-0R KNR6RB-SM-0R KNR6RB-XL-0R
77322-578EA 394.16 CAD
77322-578 77322-580 77322-582 77322-584 77322-574 77322-572 77322-570 77322-576
Men's Flame-Resistant/Chemical Splash Protection Lab Coats
Workwear
This environment of ingenuity and innovation demands reliable lab PPE that provides the ultimate in protection and comfort, for all-day wear. That’s why Bulwark CP offers breakthrough safety wear designed specifically for the lab.
  • Black lay down collar for easy recognition as a CP lab coat
  • Concealed snap front closure
  • Splash-resistant pass-throughs
  • FR only knit cuffs, safer than typical open cuff
  • Double needle top-stitched chest pocket with pencil stall
  • Two double needle top-stitched lower pockets with flaps
  • Back vent for ease of walking and sitting