NIGHTSEA® Switch Box Kit
Supplier: Electron Microscopy Sciences
NIGHTSEA®
SFA-SWK SFA-SWK
CA77463-334KT 1178.44 CAD
CA77463-334 77463-334
NIGHTSEA® Switch Box Kit
Lights
The switch box kit enables the mounting of two different wavelength excitation light heads onto a microscope, and permits switching between them easily, only needing to change the barrier filter.
  • For SFA systems only that use BNC light head connectors
  • Cannot be used with older V1 connectors
  • Includes switch box, cables, and 2 light head hangers
Specifically designed for use with NIGHTSEA® SFA stereo microscope fluorescence adapter systems that use the BNC light head connector. Users will still need to manually change the barrier filter, but it eliminates the need to swap out light heads on the gooseneck.