NIGHTSEA® Battery and Charger
Supplier: Electron Microscopy Sciences
NIGHTSEA®
SFA-BATT SFA-BATT
CA77463-304EA 970.07 CAD
77463-304 CA77463-304
NIGHTSEA® Battery and Charger
Batteries
Compact battery pack will run the NIGHTSEA® SFA stereo microscope fluorescence adapter system all day long. The battery enables truly portable operation so that users can set up demos anywhere or explore fluorescence in the field.
  • Nickel Metal Hydride (NiMH) battery
  • 8 hours continuous operation
  • 12V, 3.8 Ah capacity
  • Dimensions: 11.4×8.9×5.6 cm (4.5×3.5×2.19")
The battery enables truly portable operation, allowing demos anywhere or exploration of fluorescence in the field, without having to worry about plugging into the power grid.
The battery plugs direcly into the SFA base instead of the usual power connector. The battery has been tested at over 8 hours of continuous operation, and will last even longer if the system is turned off when not in use. Plug it into the included charger overnight to recharge.