NIGHTSEA® Tru-Block Eyeshields
Supplier: Electron Microscopy Science
SFA-EYE-C SFA-EYE-S
CA77463-310EA 74.64 CAD
CA77463-310 CA77463-312
NIGHTSEA® Tru-Block Eyeshields
Microscopes
These soft, molded rubber high-sided microscope eye shields extend far enough to truly shield the eyes from any level of ambient light and eliminate distractions for improved viewing of fluorescence subject better.
  • Standard or Compact sizes are available in pairs
  • Standard size fits 36 to 45 mm (1.45 to 1.75")
  • Compact size fits 28 to 37 mm (1.10 to 1.46")
Light entering the eyes from the side can interfere with microscopy viewing in general, and fluorescence microscopy in particular. Eye cups are available, but standard cups don't extend far from theMore Product Information